missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human THSD1 Protein, His Tag
sur devis
Spécification
Numéro d’adhésion | AAH63842 |
---|---|
Poids moléculaire | 39.7 kDa |
Source | Human THSD1, His Tag (TH1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ile 361 (Accession # AAH63842). |
Immunogène | Glu 25 - Ile 361 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15857757
|
ACROBiosystems
TH1-H52H5-100UG |
100 μg |
sur devis
100µg |
N/A | Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Cet article nécessite une mise à jour du prix avant d'être commandé. Demandez un prix pour cet article ici. |
|||||||||
15867757
|
ACROBiosystems
TH1-H52H5-1MG |
1 mg |
sur devis
1mg |
N/A | Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Cet article nécessite une mise à jour du prix avant d'être commandé. Demandez un prix pour cet article ici. |
|||||||||
Description
Thrombospondin type-1 domain-containing protein 1 (THSD1) is also known as Transmembrane molecule with thrombospondin module (TMTSP), which is a single-pass type I membrane protein and contains a type 1 thrombospondin domain that is found in thrombospondin, a number of proteins involved in the complement pathway, as well as extracellular matrix proteins. In addition, THSD1 is widely expressed on endothelial cells, with highest expression in the lung. THSD1's ligand and function are unknown, but it is postulated that THSD1 may be involved in the regulation of vasculogenesis and/or angiogenesis through its interaction with its specific ligand.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 57-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH63842 | |
Human THSD1, His Tag (TH1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ile 361 (Accession # AAH63842). | |
-20°C | |
THSD1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
39.7 kDa | |
Glu 25 - Ile 361 | |
THSD1,TMTSP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |