missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human TNFR2 / CD120b / TNFRSF1B Protein, His Tag

335.90€ - 2519.28€

Spécification

Numéro d’adhésion AAA36755
Poids moléculaire 26.0 kDa
Source Human TNFR2, His Tag (TN2-H5227) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asp 257 (Accession # AAA36755).
Immunogène Leu 23 - Asp 257
Conditions de stockage -20°C
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15867807
Afficher les documents Détails de la promotion
ACROBiosystems
TN2-H5227-50UG
50 μg
335.90€
50µg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 11-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15857807
Afficher les documents Détails de la promotion
ACROBiosystems
TN2-H5227-1MG
1 mg
2519.28€
1mg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 11-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

TNF RI is also known as the p60 or p55 TNFR) and TNF RII (the p75 or p80 TNFR) are two distinct type I transmembrane glycoproteins that bind TNF with highaffinity.Both RI and RII are prototypic members of the TNF receptor superfamily and have been designated TNFRSF1A and TNFRSF1B, respectively. Human TNF RII cDNA encodes a 461 amino acid (aa) residue precursor protein with a 22 aa putative signal peptide, a 235 aa extracellular domain, a 20 aa transmembrane domain and a 174 aa cytoplasmic domain. TNFRII is expressed in fetal brain. The protein is produced naturally as a soluble form (sTNFRII). The soluble receptor inhibits TNFα action by competing with cell surface receptors in binding TNFα, thereby blocking its biologic effects. TNFRII is strongly expressed at the cartilage–pannus junction, and plays a major role in a subset of families with multiple cases of rheumatoid arthritis (RA). Further, high plasma levels of sTNFRII were significantly associated with increased incidence of coronary heart disease, independent of established cardiovascular risk factors, and seems to be useful for monitoring the inflammatory activity of sarcoidosis.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.0 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAA36755
Human TNFR2, His Tag (TN2-H5227) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asp 257 (Accession # AAA36755).
-20°C
TNFR2
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens
26.0 kDa
Leu 23 - Asp 257
TNFRSF1B,CD120b,TBPII,TNF-R-II,TNF-R75,TNFBR,TNFR1B,TNFR2,TNFR80,p75,p75TNFR
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Vidéos
FDS
Documents

Documents

Product Certifications