missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Transthyretin / Prealbumin Protein, His Tag

Marque:  ACROBiosystems TTR-H5223-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15867877

  • 1513.19€ / 1mg
Date d’expédition estimée: 10-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Transthyretin (TTR) is also known as Prealbumin, ATTR, TBPA, PALB, which belongs to the transthyretin family. Transthyretin / TTR is a serum and cerebrospinal fluid carrier of the thyroid hormone thyroxine (T4) and retinol-binding protein bound to retinol. In cerebrospinal fluid TTR is the primary carrier of T4. TTR also acts as a carrier of retinol (vitamin A) through its association with retinol-binding protein (RBP) in the blood and the CSF. Less than 1% of TTR's T4 binding sites are occupied in blood. TTR misfolding and aggregation is known to be associated with the amyloid diseases.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 17 kDa and 33 kDa under reducing (R) condition (SDS-PAGE) due to the monomer and dimer respectively.
Spécification

Spécification

NP_000362
HEK293
Human
14.9 kDa
1 mg
Gly 21 - Glu 147
Transthyretin,TTR,Prealbumin,TBPA,ATTR,PALB,CTS,CTS1,HsT2651
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Other Recombinant Proteins
Powder
>95% as determined by SDS-PAGE
Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession # NP_000362).
-20°C
Transthyretin
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions