missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human TSLP (R127A, R130A) Protein, His Tag
423.38€ - 2951.55€
Spécification
Numéro d’adhésion | NP_149024.1 (R127A, R130A) |
---|---|
Poids moléculaire | 16.7 kDa |
Source | Human TSLP (R127A, R130A), His Tag (TSP-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1 (R127A, R130A)). |
Immunogène | Tyr 29 - Gln 159 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15877867
|
ACROBiosystems
TSP-H52HA-100UG |
100 μg |
423.38€
100µg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15887867
|
ACROBiosystems
TSP-H52HA-1MG |
1 mg |
2951.55€
1mg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Thymic stromal lymphopoietin (TSLP) is an epithelial cell-derived cytokine involved in the pathology of inflammatory skin diseases, and is widely expressed by epithelial cells. Human TSLP cDNA encodes a 159 amino acid (aa) residue precursor protein with a 28 aa signal sequence (4, 5). Human TSLP has been shown to developing nondeletional central tolerance, amplifying epithelium-induced class switching, inducing atopic diseases and maintaining intestinal noninflammatory environment. Among diverse cells responding to Human TSLP, CD11c+ dendritic cells are the most obviously characterized target cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 22-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_149024.1 (R127A, R130A) | |
Human TSLP (R127A, R130A), His Tag (TSP-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1 (R127A, R130A)). | |
-20°C | |
TSLP | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
16.7 kDa | |
Tyr 29 - Gln 159 | |
TSLP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |