missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TSPAN8 Full-length ORF (AAH05246.1, 1 a.a. - 237 a.a.) Recombinant Protein MW: 51.81kDa with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00007103-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16121442

  • 302.00€ / 10µg

Expédition estimée
31-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence: MAGVSACIKYSMFTFNFLFWLCGILILALAIWVRISNDSQAIFGSEDVGSSSYVAVDILIAVGAIIMILGFLACCGAIKESRCMLLLFFIGLLLILLLQVATGILGAVFKSKSDRIVNETLYENTKLLSATGESEKQFQEAIIVFQEEFKCCGLVNGAADWGNNFQHYPELCACLDKQRPCQSYNGKQVYKETCISFIKDFLAKNLIIVIGIAFGLAVIEILGLVFSMVLYCQIGNK
Spécification

Spécification

AAH05246.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.81kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CO-029|TM4SF3
TSPAN8
Yes
wheat germ expression system
Liquid
7103
TSPAN8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGVSACIKYSMFTFNFLFWLCGILILALAIWVRISNDSQAIFGSEDVGSSSYVAVDILIAVGAIIMILGFLACCGAIKESRCMLLLFFIGLLLILLLQVATGILGAVFKSKSDRIVNETLYENTKLLSATGESEKQFQEAIIVFQEEFKCCGLVNGAADWGNNFQHYPELCACLDKQRPCQSYNGKQVYKETCISFIKDFLAKNLIIVIGIAFGLAVIEILGLVFSMVLYCQIGNK
RUO
TSPAN8
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents