missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TXNIP Full-length ORF (NP_006463.2, 1 a.a. - 391 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
sur devis
Spécification
Nom | TXNIP (Human) Recombinant Protein (P01) |
---|---|
Recombinant | Yes |
Méthode de purification | Glutathione Sepharose 4 Fast Flow |
Poids moléculaire | 70.1kDa |
Conditions de stockage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Description
Sequence: MVMFKKIKSFEVVFNDPEKVYGSGEKVAGRVIVEVCEVTRVKAVRILACGVAKVLWMQGSQQCKQTSEYLRYEDTLLLEDQPTGENEMVIMRPGNKYEYKFGFELPQGPLGTSFKGKYGCVDYWVKAFLDRPSQPTQETKKNFEVVDLVDVNTPDLMAPVSAKKEKKVSCMFIPDGRVSVSARIDRKGFCEGDEISIHADFENTCSRIVVPKAAIVARHTYLANGQTKVLTQKLSSVRGNHIISGTCASWRGKSLRVQKIRPSILGCNILRVEYSLLIYVSVPGSKKVILDLPLVIGSRSGLSSRTSSMASRTSSEMSWVDLNIPDTPEAPPCYMDVIPEDHRLESPTTPLLDDMDGSQDSPIFMYAPEFKFMPPPTYTEVDPCILNNNVQSpécification
TXNIP (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
70.1kDa |