missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UBE2Q2 Full-length ORF (NP_775740.1, 1 a.a. - 375 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00092912-P01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16126783

  • 508.00€ / 25µg
Expédition estimée: 07-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence: MSVSGLKAELKFLASIFDKNHERFRIVSWKLDELHCQFLVPQQGSPHSLPPPLTLHCNITESYPSSSPIWFVDSEDPNLTSVLERLEDTKNNNLLRQQLKWLICELCSLYNLPKHLDVEMLDQPLPTGQNGTTEEVTSEEEEEEEEMAEDIEDLDHYEMKEEEPISGKKSEDEGIEKENLAILEKIRKTQRQDHLNGAVSGSVQASDRLMKELRDIYRSQSYKTGIYSVELINDSLYDWHVKLQKVDPDSPLHSDLQILKEKEGIEYILLNFSFKDNFPFDPPFVRVVLPVLSGGYVLGGGALCMELLTKQGWSSAYSIESVIMQINATLVKGKARVQFGANKNQYNLARAQQSYNSIVQIHEKNGWYTPPKEDG
Spécification

Spécification

NP_775740.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
69.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp762C143
UBE2Q2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
92912
UBE2Q2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSVSGLKAELKFLASIFDKNHERFRIVSWKLDELHCQFLVPQQGSPHSLPPPLTLHCNITESYPSSSPIWFVDSEDPNLTSVLERLEDTKNNNLLRQQLKWLICELCSLYNLPKHLDVEMLDQPLPTGQNGTTEEVTSEEEEEEEEMAEDIEDLDHYEMKEEEPISGKKSEDEGIEKENLAILEKIRKTQRQDHLNGAVSGSVQASDRLMKELRDIYRSQSYKTGIYSVELINDSLYDWHVKLQKVDPDSPLHSDLQILKEKEGIEYILLNFSFKDNFPFDPPFVRVVLPVLSGGYVLGGGALCMELLTKQGWSSAYSIESVIMQINATLVKGKARVQFGANKNQYNLARAQQSYNSIVQIHEKNGWYTPPKEDG
RUO
UBE2Q2
Wheat Germ (in vitro)
GST
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions