Learn More
ACROBiosystems Human UCH-L1 Protein, His Tag
302.42€ - 1838.68€
Spécification
Numéro d’adhésion | NP_004172 |
---|---|
Poids moléculaire | 25.7 kDa |
Source | Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Gln 2 - Ala 223 (Accession # NP_004172). |
Immunogène | Gln 2 - Ala 223 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15867887
|
ACROBiosystems
UC1-H5140-50UG |
50 μg |
302.42€
50µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15857887
|
ACROBiosystems
UC1-H5140-1MG |
1 mg |
1838.68€
1mg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Ubiquitin carboxyl-terminal hydrolase isozyme L1 (UCHL1) ,a member of the peptidase C12 family, is also known as neuron cytoplasmic protein 9.5 (PGP 9.5) and ubiquitin thioesterase L1.About 30% of total UCHL1 is associated with membranes in brain. UCHL1 is involved both in the processing of ubiquitin precursors and of ubiquitinated proteins. This enzyme is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. Also,UCHL1 can bind to free monoubiquitin and may prevent its degradation in lysosomes. The homodimer of UCHL1 may have ATP-independent ubiquitin ligase activity. Furthermore, the mutations of UCHL1 gene can result in parkinson disease 5.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.7 kDa. The protein migrates as 27-28 kDa under reducing (R) condition (SDS-PAGE).Spécification
NP_004172 | |
Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Gln 2 - Ala 223 (Accession # NP_004172). | |
-20°C | |
UCH-L1 | |
Human | |
50 mM Tris, 150 mM Nacl, pH8.0 | |
E.coli | |
Other Recombinant Proteins |
25.7 kDa | |
Gln 2 - Ala 223 | |
UCHL1,PGP9.5 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |