Learn More
ACROBiosystems Human VCAM-1 / CD106 Protein, His Tag
302.42€ - 2020.12€
Spécification
Numéro d’adhésion | NP_001069 |
---|---|
Poids moléculaire | 74.9 kDa |
Source | Human VCAM-1, His Tag (VC1-H5224) is expressed from human 293 cells (HEK293). It contains AA Phe 25 - Pro 697 (Accession # NP_001069). |
Immunogène | Phe 25 - Pro 697 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15857897
|
ACROBiosystems
VC1-H5224-100UG |
100 μg |
302.42€
100µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15867897
|
ACROBiosystems
VC1-H5224-1MG |
1 mg |
2020.12€
1mg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Vascular cell adhesion protein 1 (VCAM1) is also known as CD106, INCAM-100 and L1CAM, is a cell surface sialoglycoprotein belonging to the immunoglobulin superfamily. VCAM1 / CD106 contains 7 Ig-like C2-type (immunoglobulin-like) domains. CD106 / VCAM-1 is expressed on inflammed vascular endothelium, as well as on macrophage-like and dendritic cell types in both normal and inflammed tissue. L1CAM / VCAM-1 is Important in cell-cell recognition and appears to function in leukocyte-endothelial cell adhesion. CD106 / VCAM1 interacts with the beta-1 integrin VLA4 on leukocytes, and mediates both adhesion and signal transduction. The VCAM1 / VLA4 interaction may play a pathophysiologic role both in immune responses and in leukocyte emigration to sites of inflammation. INCAM-100 / VCAM1 binds to ECMV-D capsid proteins and acts as a receptor for this virus.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 74.9 kDa. The protein migrates as 80-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_001069 | |
Human VCAM-1, His Tag (VC1-H5224) is expressed from human 293 cells (HEK293). It contains AA Phe 25 - Pro 697 (Accession # NP_001069). | |
-20°C | |
VCAM-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
74.9 kDa | |
Phe 25 - Pro 697 | |
VCAM1,CD106,INCAM-100,V-CAM 1,VCAM-1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |