missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human VEGF R1 / Flt-1 Protein, His Tag

Marque:  ACROBiosystems VE1-H5220-100UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15867907

  • 292.55€ / 100µg
Date d’expédition estimée: 08-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Vascular endothelial growth factor receptor 1 (VEGFR1) is also known as Fms-like tyrosine kinase 1 (FLT-1), Tyrosine-protein kinase receptor FLT, is a single-pass type I membrane protein and secreted protein which belongs to the protein kinase superfamily, Tyr protein kinase family and CSF-1/PDGF receptor subfamily. VEGFR1 is detected in normal lung, but also in placenta, liver, kidney, heart and brain tissues and specifically expressed in most of the vascular endothelial cells, and also expressed in peripheral blood monocytes. VEGFR1 acts as a cell-surface receptor for VEGFA, VEGFB and PGF, and plays an essential role in the development of embryonic vasculature, the regulation of angiogenesis, cell survival, cell migration, macrophage function, chemotaxis, and cancer cell invasion. VEGFR1 may play an essential role as a negative regulator of embryonic angiogenesis by inhibiting excessive proliferation of endothelial cells. VEGFR1 can promote endothelial cell proliferation, survival and angiogenesis in adulthood.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 83.3 kDa. The protein migrates as 110-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

NP_002010.1
HEK293
Human
83.3 kDa
100 µg
Ser 27 - Asn 756
FLT,VEGFR1,FLT1
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human VEGF R1, His Tag (VE1-H5220) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Asn 756 (Accession # NP_002010.1).
-20°C
VEGF R1
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions