missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human VEGF R3 / FLT4 Protein, Fc Tag

Marque:  ACROBiosystems FL4-H5251-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15866767

  • 1997.41€ / 1mg
Date d’expédition estimée: 07-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Vascular endothelial growth factor receptor 3 (VEGF R3), also known as FLT-4, together with the other two members VEGFR1 (FLT-1) and VEGFR2 (KDR/Flk-1) are receptors for vascular endothelial growth factors (VEGF) and belong to the class III subfamily of receptor tyrosine kinases (RTKs). VEGF R3 mediates lymphangiogenesis in response to VEGF-C and VEGF-D. VEGF R3 is widely expressed in the early embryo but becomes restricted to lymphatic endothelia at later stages of development. It is likely that VEGF R3 may be important for lymph angiogenesis.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 111.2 kDa. As a result of glycosylation, the protein migrates as 140-160 kDa under reducing (R) condition, and 280-320 kDa under non-reducing (NR) condition (SD
Spécification

Spécification

NP_002011.2
HEK293
Human
111.2 kDa
1 mg
Tyr 25 - Ile 776
Flt-4,FLT4,LMPH1A,PCLFLT41,VEGFR3,VEGFR-3,FLT-4,FLT-41,FLT41,PCL
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human VEGF R3, Fc Tag (FL4-H5251) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Accession # NP_002011.2).
-20°C
VEGF R3
Fc Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions