Learn More
ACROBiosystems Human VSIG8 Protein, His Tag
Marque: ACROBiosystems VS8-H52H1-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
V-set and immunoglobulin domain containing 8 (VSIG8), also known as C1orf204, is a type I transmembrane protein of the B7 family within the Ig superfamily. VSIG8 was identified from proteomic analysis of human hair shafts. It is expressed in the hair follicle and shaft, superficial layers of the nail matrix, and superficial layers of oral epithelium.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.0 kDa. The protein migrates as 30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_001013683.1 | |
1 mg | |
Val 22 - Gly 263 | |
VSIG8,C1orf204 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
29.0 kDa | |
Human VSIG8, His Tag (VS8-H52H1) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession # NP_001013683.1). | |
-20°C | |
VSIG8 | |
Human | |
20 mM NaAC, 150 mM NaCl, pH4.0 | |
HEK293 | |
Other Recombinant Proteins |