missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF274 Full-length ORF (NP_598009.1, 1 a.a. - 653 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Spécification
Numéro d’adhésion | NP_598009.1 |
---|---|
À utiliser avec (application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 10782 |
Poids moléculaire | 100.6kDa |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16125692
|
Abnova™
H00010782-P01.25UG |
25 ug |
508.00€
25µg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16115692
|
Abnova™
H00010782-P01.10UG |
10 ug |
335.00€
10µg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene encodes a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The encoded protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus. Alternatively spliced transcript variants encoding different isoforms exist. These variants utilize alternative polyadenylation signals. [provided by RefSeq]
Sequence: MASRLPTAWSCEPVTFEDVTLGFTPEEWGLLDLKQKSLYREVMLENYRNLVSVEHQLSKPDVVSQLEEAEDFWPVERGIPQDTIPEYPELQLDPKLDPLPAESPLMNIEVVEVLTLNQEVAGPRNAQIQALYAEDGSLSADAPSEQVQQQGKHPGDPEAARQRFRQFRYKDMTGPREALDQLRELCHQWLQPKARSKEQILELLVLEQFLGALPVKLRTWVESQHPENCQEVVALVEGVTWMSEEEVLPAGQPAEGTTCCLEVTAQQEEKQEDAAICPVTVLPEEPVTFQDVAVDFSREEWGLLGPTQRTEYRDVMLETFGHLVSVGWETTLENKELAPNSDIPEEEPAPSLKVQESSRDCALSSTLEDTLQGGVQEVQDTVLKQMESAQEKDLPQKKHFDNRESQANSGALDTNQVSLQKIDNPESQANSGALDTNQVLLHKIPPRKRLRKRDSQVKSMKHNSRVKIHQKSCERQKAKEGNGCRKTFSRSTKQITFIRIHKGSQVCRCSECGKIFRNPRYFSVHKKIHTGERPYVCQDCGKGFVQSSSLTQHQRVHSGERPFECQECGRTFNDRSAISQHLRTHTGAKPYKCQDCGKAFRQSSHLIRHQRTHTGERPYACNKCGKAFTQSSHLIGHQRTHNRTKRKKKQPTSSpécification
NP_598009.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
100.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MASRLPTAWSCEPVTFEDVTLGFTPEEWGLLDLKQKSLYREVMLENYRNLVSVEHQLSKPDVVSQLEEAEDFWPVERGIPQDTIPEYPELQLDPKLDPLPAESPLMNIEVVEVLTLNQEVAGPRNAQIQALYAEDGSLSADAPSEQVQQQGKHPGDPEAARQRFRQFRYKDMTGPREALDQLRELCHQWLQPKARSKEQILELLVLEQFLGALPVKLRTWVESQHPENCQEVVALVEGVTWMSEEEVLPAGQPAEGTTCCLEVTAQQEEKQEDAAICPVTVLPEEPVTFQDVAVDFSREEWGLLGPTQRTEYRDVMLETFGHLVSVGWETTLENKELAPNSDIPEEEPAPSLKVQESSRDCALSSTLEDTLQGGVQEVQDTVLKQMESAQEKDLPQKKHFDNRESQANSGALDTNQVSLQKIDNPESQANSGALDTNQVLLHKIPPRKRLRKRDSQVKSMKHNSRVKIHQKSCERQKAKEGNGCRKTFSRSTKQITFIRIHKGSQVCRCSECGKIFRNPRYFSVHKKIHTGERPYVCQDCGKGFVQSSSLTQHQRVHSGERPFECQECGRTFNDRSAISQHLRTHTGAKPYKCQDCGKAFRQSSHLIRHQRTHTGERPYACNKCGKAFTQSSHLIGHQRTHNRTKRKKKQPTS | |
RUO | |
ZNF274 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10782 | |
ZNF274 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686K08243/FLJ37843/HFB101/ZF2/ZKSCAN19 | |
ZNF274 | |
Recombinant | |
wheat germ expression system |