Learn More
Novus Biologicals™ Human Zonulin ELISA Kit (Colorimetric)
Marque:  Novus Biologicals™ NBP3-21146
Code produit. 18650164
  • 967.00€ / 1 kit
Description
TRUSTED_SUSTAINABILITY
Spécification
Spécification
0.625 - 40 ng/mL
ELISA Kit (Colorimetric)
This assay has high sensitivity and excellent specificity for detection of human zonulin. No significant cross-reactivity or interference between human zonulin and analogues was observed.
1 kit
Human
0.156 ng/mL
Plasma, Serum, Tissue Homogenates
ELISA
Store at 4°C.