missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOSLG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14922-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
CD275 is a ligand for the T-cell-specific cell surface receptor ICOS. Acts as a costimulatory signal for T-cell proliferation and cytokine secretion; induces also B-cell proliferation and differentiation into plasma cells. Could play an important role in mediating local tissue responses to inflammatory conditions, as well as in modulating the secondary immune response by co-stimulating memory T-cell function.Spécification
ICOSLG | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75144 | |
B7 H2, B7 homolog 2, B7 like protein Gl50, B7 related protein 1, B7H2, B7RP 1, B7RP1, CD275, GL50, ICOS L, ICOS ligand, ICOSL, ICOSLG, KIAA0653, LICOS | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
23308 |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
ICOSLG | |
Liquid | |
ICOSLG | |
ICOSLG Fusion Protein Ag6392 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |