missing translation for 'onlineSavingsMsg'
Learn More

IgD Rat anti-Mouse, APC-eFluor® 780, Clone: 11-26c (11-26), eBioscience™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 47-5993-80

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15346128

  • 122.00€ / 25µg

Expédition estimée
26-10-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
Spécification

Spécification

IgD
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
11-26c (11-26)
APC-eFluor™ 780
Ighd
Ighd
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
380797
Vidéos
Documents

For Research Use Only.