Learn More
IgD Mouse anti-Human, Super Bright 436, Clone: IA6-2, eBioscience™
Mouse Monoclonal Antibody
Marque: Invitrogen 62-9868-42
Informations supplémentaires : Poids : 0.01000kg
Description
Description: This IA6-2 monoclonal antibody reacts with the heavy chain of human immunoglobulin D (IgD). This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding. Applications Reported: This IA6-2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This IA6-2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.
Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.Spécification
IgD | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
IGHD | |
Affinity chromatography | |
RUO | |
3495 | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
IA6-2 | |
Super Bright 436 | |
IGHD | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Liquid |