missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), eFluor™ 660, eBioscience™, Invitrogen™

Mouse Isotype Control

Marque:  Affymetrix eBioscience 50-4714-82

182.00 EUR valable jusqu'au 2024-03-29
Utilisez le code promo "21615" pour bénéficier de cette offre.



Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.25000kg

 Afficher plus de versions de ce produit

Code produit. 15331910

  • 273.00€ / 100µg
En stock
pour voir le stock disponible 
Ajouter au panier

Description

Description

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: The Mouse IgG1 K Isotype Control has been reported for use in flow cytometric analysis and immunohistochemical staining. Applications Tested: The Mouse IgG1 K Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at the same concentration as the experimental antibody. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunohistochemistry.
Spécification

Spécification

Mouse IgG1 kappa
P3.6.2.8.1
eFluor 660
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry, Immunohistochemistry
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Mouse
100 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG1 κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.