missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGJ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13688-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
J chain is a small glycopeptide and is structurally unrelated to heavy or light chains, but is synthesized by all plasma cells that secrete polymeric immunoglobulins. J chain is linked to IgA and IgM by disulfide bonds, however, it has been detected in IgG- and IgD-containing cells. J chains are present in a large proportion of the immunoglobulin-positive cells in the germinal centers of the tonsil and lymph node. B cells secrete J chain at an early stage of differentiation with the expression persisting in those cells destined to produce IgA or IgM.Spécification
IGJ | |
Polyclonal | |
Unconjugated | |
JCHAIN | |
IGCJ, IGJ, Immunoglobulin J chain, JCH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16069, 3512, 360922 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01591, P01592 | |
JCHAIN | |
IGJ Fusion Protein Ag4559 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |