Learn More
IL-10 Rat anti-Mouse, APC, Clone: JES5-16E3, BD
Rat Monoclonal Antibody
175.00€ - 428.00€
Spécification
Antigène | IL-10 |
---|---|
Clone | JES5-16E3 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15870729
|
BD Biosciences
561059 |
25 μg |
175.00€
1 pièce |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15827718
|
BD Biosciences
554468 |
0.1 mg |
428.00€
1 pièce |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The JES5-16E3 monoclonal antibody specifically binds to the mouse cytokine, Interleukin-10 (IL-10). IL-10 is also known as Cytokine Synthesis Inhibitory Factor (CSIF). It is produced by various activated cell types including CD4+ T cells, CD8+ T cells, T regulatory cells, NK T cells, B1 B cells, NK cells, macrophages, dendritic cells, mast cells, granulocytes and keratinocytes. IL-10 plays a pivotal role in regulating immune responses and protecting the host from damage caused by inflammatory and autoimmune responses. IL-10 has numerous biological activities including the inhibition of cytokine synthesis by activated T cells, NK cells, monocytes, and macrophages. In the presence of accessory cells, IL-10 inhibits mitogen- or anti-CD3 induced proliferation of T lymphocytes. IL-10 has also been shown to costimulate the development of thymocytes, B cell differentiation and the generation of cytotoxic T cells. The immunogen used to generate the JES5-16E3 hybridoma was recombinant mouse IL-10. JES5-16E3 is a neutralizing antibody.
Spécification
IL-10 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Recombinant mouse IL-10 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
JES5-16E3 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2b | |
Affinity Purified |
For Research Use Only.