missing translation for 'onlineSavingsMsg'
Learn More

IL-21 Rat anti-Mouse, eFluor® 660, Clone: FFA21, eBioscience™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 50-7211-80

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.25000kg

 Afficher plus de versions de ce produit

Code produit. 15341950

  • sur devis / 25µg

Pour délai, nous consulter 03 88 67 14 14

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Cet article ne peut pas être commandé en ligne car le prix de vente peut varier. Pour en savoir plus, veuillez contacter notre service client au 03 88 67 14 14 .
Voir les produits de remplacement »


Description

Description

Applications Tested: This FFA21 antibody has been tested by intracellular staining and flow cytmetric analysis of Th17 stimulated mouse cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintece by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.
Spécification

Spécification

IL-21
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9ES17
Interleukin-21, IL-21, Za11
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
FFA21
eFluor™ 660
Il21
Liquid
Il21
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
60505
Vidéos
FDS
Documents

For Research Use Only.