Learn More
IL-33R (ST2) Monoclonal Antibody (hIL33Rcap), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
175.00€ - 382.00€
Spécification
Antigène | IL-33R (ST2) |
---|---|
Clone | hIL33Rcap |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16370734
|
Affymetrix eBioscience
17-9338-41 |
25 Tests |
175.00€
25 pièces |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16380734
|
Affymetrix eBioscience
17-9338-42 |
100 Tests |
382.00€
100 pièces |
En stock Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Description: This hIL33Rcap monoclonal antibody recognizes human IL-33R, also known as ST2. This antibody binds both, the membrane bound form (ST2L) and the soluble form (ST2S). This antibody is suitable for both surface and intracellular staining. This hIL33Rcap clone is also used as a capture antibody in the IL-33R (ST2) Human ProcartaPlex™ Simplex Kit (Product # EPX010-12231-901) Applications Reported: This hIL33Rcap antibody has been reported for use in flow cytometric analysis. Applications Tested: This hIL33Rcap antibody has been pre-diluted and tested by flow cytometric analysis of LAD2 cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
IL-33R (ST2) Monoclonal antibody specifically detects IL-33R (ST2) in Human samples. It is validated for Flow Cytometry.Spécification
IL-33R (ST2) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; IL1RL1; IL-1RL1; IL33R; IL-33R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2 | |
IL1RL1 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
hIL33Rcap | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
Q01638 | |
9173 | |
Extracellular domain of human IL-33R protein | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL1RL1 |