missing translation for 'onlineSavingsMsg'
Learn More

IL-33R (ST2) Mouse anti-Human, APC, Clone: hIL33Rcap, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Marque:  Invitrogen 17-9338-41

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16370734

  • 140.00€ / 25 pièces

Expédition estimée
16-11-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

IL-33R (ST2) Monoclonal antibody specifically detects IL-33R (ST2) in Human samples. It is validated for Flow Cytometry
Spécification

Spécification

IL-33R (ST2)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q01638
DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; IL-1 R4; Il1rl1; IL-1RL1; IL33R; IL-33R; Interleukin1 receptor like 1; Interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; Lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2
Mouse
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
9173
Flow Cytometry
hIL33Rcap
APC
IL1RL1
Liquid
IL1RL1
Extracellular domain of human IL-33R protein
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Vidéos
Documents