missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL10RB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | IL10RB |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16814124
|
Proteintech
15102-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 03-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16804124
|
Proteintech
15102-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 22-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.Spécification
IL10RB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q08334, Q61190 | |
16155, 3588 | |
IL10RB Fusion Protein Ag7445 | |
Primary | |
-20°C | |
IL10RB |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CDW210B, CRF2 4, CRFB4, D21S58, D21S66, IL 10 receptor subunit beta, IL 10R subunit 2, IL 10R subunit beta, IL 10R2, IL 10RB, IL10RB, interleukin 10 receptor, beta | |
IL10RB | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |