missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL17RC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15648-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a single-pass type I membrane protein that shares similarity with the interleukin-17 receptor (IL-17RA). Unlike IL-17RA, which is predomitly expressed in hemopoietic cells, and binds with high affinity to only IL-17A, this protein is expressed in nonhemopoietic tissues, and binds both IL-17A and IL-17F with similar affinities. The proinflammatory cytokines, IL-17A and IL-17F, have been implicated in the progression of inflammatory and autoimmune diseases. Multiple alternatively spliced transcript variants encoding different isoforms have been detected for this gene, and it has been proposed that soluble, secreted proteins lacking transmembrane and intracellular domains may function as extracellular antagonists to cytokine signaling.Spécification
IL17RC | |
Polyclonal | |
Unconjugated | |
IL17RC | |
IL 17 receptor C, IL 17RC, IL 17RL, IL17 RL, IL17RC, IL17Rhom, IL17RL, interleukin 17 receptor C, ZcytoR14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171095, 84818 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K4C2, Q8NAC3 | |
Il17rc | |
IL17RC Fusion Protein Ag8190 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |