missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1RAPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | IL1RAPL1 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16811215
|
Proteintech
21609-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 03-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16801215
|
Proteintech
21609-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 03-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The protein encoded by this gene is a member of the interleukin 1 receptor family and is similar to the interleukin 1 accessory proteins. This protein has an N-terminal signal peptide, three extracellular immunoglobulin Ig-like domains, a transmembrane domain, an intracellular Toll/IL-1R domain, and a long C-terminal tail which interacts with multiple signalling molecules. This gene is located at a region on chromosome X that is associated with a non-syndromic form of X-linked intellectual disability. Deletions and mutations in this gene were found in patients with intellectual disability. This gene is expressed at a high level in post-natal brain structures involved in the hippocampal memory system, which suggests a specialized role in the physiological processes underlying memory and learning abilities, and plays a role in synapse formation and stabilization.Spécification
IL1RAPL1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P59823, P59824, Q9NZN1 | |
11141, 317553, 331461 | |
IL1RAPL1 Fusion Protein Ag16252 | |
Primary | |
-20°C | |
IL1RAPL1 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
IL 1 RAPL 1, IL 1RAPL 1, IL1R8, IL1RAPL, IL1RAPL 1, IL1RAPL1, MRX10, MRX21, MRX34, Oligophrenin 4, OPHN4, TIGIRR 2 | |
Il1rapl1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |