missing translation for 'onlineSavingsMsg'
Learn More

IL1RAPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 21609-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16801215

  • 329.85€ / 150µL

Expédition estimée
08-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a member of the interleukin 1 receptor family and is similar to the interleukin 1 accessory proteins. This protein has an N-terminal signal peptide, three extracellular immunoglobulin Ig-like domains, a transmembrane domain, an intracellular Toll/IL-1R domain, and a long C-terminal tail which interacts with multiple signalling molecules. This gene is located at a region on chromosome X that is associated with a non-syndromic form of X-linked intellectual disability. Deletions and mutations in this gene were found in patients with intellectual disability. This gene is expressed at a high level in post-natal brain structures involved in the hippocampal memory system, which suggests a specialized role in the physiological processes underlying memory and learning abilities, and plays a role in synapse formation and stabilization.
Spécification

Spécification

IL1RAPL1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P59823, P59824, Q9NZN1
IL 1 RAPL 1, IL 1RAPL 1, IL1R8, IL1RAPL, IL1RAPL 1, IL1RAPL1, MRX10, MRX21, MRX34, Oligophrenin 4, OPHN4, TIGIRR 2
Rabbit
IgG
150 μL
-20°C
Primary
11141, 317553, 331461
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
IL1RAPL1
Liquid
Il1rapl1
IL1RAPL1 Fusion Protein Ag16252
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents