missing translation for 'onlineSavingsMsg'
Learn More

IL1RL1 Recombinant Human Protein, His Tag

IL1RL1 Recombinant Human Protein, His Tag

Marque:  Invitrogen 10105H08H250

Code nomenclature Nacres: NA.47

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 13472929

  • 614.00€ / 250µg


Date dexpédition estimée
du fournisseur 04-07-2019
Ajouter au panier

Description

Description

Interleukin 1 receptor-like 1 (IL1RL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human IL1RL1, isoform 2 (NP_003847.2), residues 1 to 328, fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA. Immobilized recombinant human IL33 at 10μg/ml(100μl/well) can bind biotinylated human IL1RL1 with a linear range of 0.16-8μg/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

Spécification

Spécification

Protein assays and analysis
Lyophilized
36.5kDa
≥90% by SDS-PAGE
–20 to –70°C
5 Pack(s)
IL1RL1, T1, ST2, DER4, ST2L, ST2V, FIT-1, MGC32623
IL1RL1
Cytokines and receptors
IL1RL1 Recombinant Human Protein
His Tag
Lyophilized
9173
IL1RL1 Recombinant Human Protein
5 x 50μg
For research use only. Not for use in diagnostic procedures
Cytokine receptors, interleukins
IL1RL1 Recombinant Human Protein
Assay standard
Mammalian
Human
Vidéos

For Research Use Only. Not for use in diagnostic procedures.