missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMMP2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15970-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
IMP2L complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 are the catalytic subunits of the IMP complex.Spécification
IMMP2L | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BPT6, Q96T52 | |
IMMP2L, IMP2, IMP2 LIKE, IMP2 like protein | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
100359529, 83943, 93757 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
IMMP2L | |
Liquid | |
Immp2l | |
IMMP2L Fusion Protein Ag8752 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |