missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Mouse anti-Human, Mouse, Clone: 4B6A7, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | Insulin |
---|---|
Clone | 4B6A7 |
Concentration | 1.23 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16847855
|
Proteintech
66198-1-IG-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16837855
|
Proteintech
66198-1-IG-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.Spécification
Insulin | |
1.23 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ILPR, INS, Insulin, IRDN | |
INS | |
IgG2a | |
Protein A | |
Antibody |
4B6A7 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P01308 | |
3630 | |
Insulin Fusion Protein Ag8630 | |
Primary | |
-20°C | |
INS |