missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAB1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27511-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the "Signalosome", which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.Spécification
JAB1 | |
Polyclonal | |
Unconjugated | |
COPS5 | |
COPS5, CSN5, JAB1, MOV 34, SGN5, Signalosome subunit 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10987 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q92905 | |
COPS5 | |
JAB1 Fusion Protein Ag25950 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |