missing translation for 'onlineSavingsMsg'
Learn More

KANK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 21733-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16801335

  • 329.85€ / 150µL

Expédition estimée
10-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the KN motif and ankyrin repeat domains (KANK) family of proteins, which play a role in cytoskeletal formation by regulating actin polymerization. The encoded protein functions in the sequestration of steroid receptor coactivators and possibly other proteins. Mutations in this gene are associated with impaired kidney podocyte function and nephrotic syndrome, and keratoderma and woolly hair.
Spécification

Spécification

KANK2
0.46 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q63ZY3, Q8BX02
ANKRD25, KANK2, KIAA1518, MXRA3, SIP, SRC 1 interacting protein, SRC1 interacting protein
Rabbit
IgG
150 μL
-20°C
Primary
100361376, 235041, 25959
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
KANK2
Liquid
Kank2
KANK2 Fusion Protein Ag16416
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents