Learn More
KCNJ14 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14171-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16832694
  • 121.38€ / 20µL
Expédition estimée: 29-09-2022
pour voir le stock disponible 
Description
Description
The KCNJ14 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human KCNJ14. This antibody recognizes human, mouse, rat antigen. The KCNJ14 antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
Spécification
KCNJ14
Polyclonal
Unconjugated
20 μL
3770
Liquid
ELISA, Western Blot
467 μg/mL
Rabbit
Primary
Rat, Mouse, Human