missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM5D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22739-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
EXOSC3 is component of the exosome 3′->5′ exoribonuclease complex and is required for the 3′ processing of the 7S pre-RNA to the mature 5.8S rRNA.Spécification
KDM5D | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BY66 | |
H Y, Histocompatibility Y antigen, Histone demethylase JARID1D, HY, HYA, JARID1D, KDM5D, KIAA0234, lysine (K) demethylase 5D, Lysine specific demethylase 5D, Protein SmcY, SMCY | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
8284 |
Western Blot | |
Unconjugated | |
KDM5D | |
Liquid | |
KDM5D | |
KDM5D Fusion Protein Ag18690 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |