missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15681-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified.Spécification
KHK | |
Polyclonal | |
Unconjugated | |
KHK | |
Hepatic fructokinase, Ketohexokinase, ketohexokinase (fructokinase), KHK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16548, 25659, 3795 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P50053, P97328, Q02974 | |
Khk | |
KHK Fusion Protein Ag8182 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |