missing translation for 'onlineSavingsMsg'
Learn More

Ki-67 Monoclonal Antibody (SolA15), APC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

165.00€ - 406.00€

Specifica

Antigène Ki-67
Clone SolA15
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantité Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantité Prezzo Quantità e disponibilità  
15875353
missing translation for 'viewDocuments' Dettagli della promozione
Affymetrix eBioscience
17-5698-80
25 μg
165.00€
25µg
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Expédition estimée: 16-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15885353
missing translation for 'viewDocuments' Dettagli della promozione
Affymetrix eBioscience
17-5698-82
100 μg
406.00€
100µg
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
En stock
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Descrizione

Descrizione

Description: The monoclonal antibody SolA15 recognizes mouse and rat Ki-67, a 300 kDa nuclear protein. Ki-67 is present during all active phases of the cell cycle (G1, S, G2, and mitosis), but is absent from resting cells (G0). Ki-67 is detected within the nucleus during interphase but redistributes to the chromosomes during mitosis. Ki-67 is used as a marker for determining the growth fraction of a given population of cells. In studies of tumor cells, the "Ki-67 labeling index" refers to the number of Ki-67 positive cells within the population and this is used to predict outcome of particular cancer types. Ki-67 has been shown to interact with the DNA-bound protein chromobox protein homolog 3 (CBX3) (heterochromatin). The SolA15 antibody also recognizes human, non-human primate and canine Ki-67. Applications Reported: This SolA15 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This SolA15 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular proteins located under the Resources Tab online. This antibody can be used at less than or equal to 0.06 μg per test.

Ki-67 Monoclonal antibody specifically detects Ki-67 in Canine, Cynomolgus Monkey, Human, Mouse, Non-human Primate, Rat samples. It is validated for Flow Cytometry.
Specifica

Specifica

Ki-67
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
antigen identified by monoclonal antibody Ki 67; antigen identified by monoclonal antibody Ki-67; Antigen identified by monoclonal antibody Ki-67 homolog; Antigen KI-67; Antigen KI-67 homolog; antigen KI-67; proliferation marker protein Ki-67; antigen KI-67-like; cb31; D630048A14Rik; I79_022666; Ki67; Ki-67; KIA; LOW QUALITY PROTEIN: proliferation marker protein Ki-67; marker of proliferation Ki-67; MIB-; MIB-1; Mki67; PPP1R105; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; sb:cb31; si:ch211-250b22.7; unnamed protein product; wu:fa11g09; wu:fb57a07; wu:fi14e05
Mki67
Primary
4° C, store in dark, DO NOT FREEZE!
Mki67
SolA15
Flow Cytometry
APC
Rat
Canine, Cynomolgus Monkey, Human, Mouse, Non-human Primate, Rat
E9PVX6, P46013
100686578, 102135895, 17345, 291234, 4288
IgG2a κ
Affinity chromatography
Antibody
Videos
SDS
Documenti

Documenti

Certificati