missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0368 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25684-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Ecm29 appears to be one of three proteins which associate significantly with proteasomes. Ecm29 has been shown to enhance the stability of the proteasome. Ecm29 deletion has been shown to produce 26S proteasomes which dissociated in the absence of ATP. It was thus suggested that Ecm29 tethers the regulatory complex to the proteasome.Spécification
KIAA0368 | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5VYK3 | |
ECM29, extracellular mutant 29, KIAA0368, KIAA1962, RP11 386D8.2 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
23392 |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
ECPAS | |
Liquid | |
ECPAS | |
KIAA0368 Fusion Protein Ag22309 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |