missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0774 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24921-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
CAZIP, also known as TIP150, contains a leucine-zipper domain at the C-terminus and has at least three isoforms. CAZIP is expressed in heart during early cardiac development and in parts of the nervous system in later embryonic development. It is thought to bind microtubules and facilitate the end-binding protein 1 (EB1)-dependent loading of MCAK onto microtubules plus ends and orchestrates the dynamics at the plus end of microtubules. CAZIP may regulate the dynamics of microtubules at their growing distal tip.Spécification
KIAA0774 | |
Polyclonal | |
Unconjugated | |
MTUS2 | |
Cardiac zipper protein, CAZIP, KIAA0774, MTUS2, TIP150, Tracking protein of 150 kDa | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23281, 77521 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3UHD3, Q5JR59 | |
Mtus2 | |
KIAA0774 Fusion Protein Ag20593 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |