Learn More
KIAA1797 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 30140-1-AP-20UL
Beskrivning
The KIAA1797 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIAA1797. This antibody recognizes human, mouse, rat antigen. The KIAA1797 antibody has been validated for the following applications: WB, ELISA analysis.
Specifikationer
| KIAA1797 | |
| 850 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| FLJ20375, KIAA1797 | |
| 20 μL | |
| 54914 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.