missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27218-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Kif2a gene ontology annotations related to this gene include microtubule-based movement.Spécification
KIF2A | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00139, P28740 | |
HK2, KIF2, KIF2A, Kinesin 2, kinesin heavy chain member 2A, Kinesin like protein KIF2A, KNS2 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
16563, 3796 |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
KIF2A | |
Liquid | |
KIF2A | |
KIF2A Fusion Protein Ag25692 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human |