Learn More
KIF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17290-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
KIF6 belongs to the kinesin-like protein family. There are four named isoforms. The KIF6 719Arg variant has been associated with an increased risk of coronary disease and an increased response to statin therapy.Spécification
KIF6 | |
Polyclonal | |
Unconjugated | |
KIF6 | |
C6orf102, KIF6, kinesin family member 6, Kinesin like protein KIF6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171291, 221458, 319991 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZMV9 | |
Kif6 | |
KIF6 Fusion Protein Ag11164 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |