missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir7.1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12657-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. KCNJ13 has a very low single channel conductance, low sensitivity to block by external barium and cesium, and no dependence of its inward rectification properties on the internal blocking particle magnesium.Spécification
Kir7.1 | |
Polyclonal | |
Unconjugated | |
KCNJ13 | |
KCNJ13, KIR1.4, Kir7.1, SVD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3769 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60928 | |
KCNJ13 | |
Kir7.1 Fusion Protein Ag3327 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |