missing translation for 'onlineSavingsMsg'
Learn More

KRAP/SSFA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14157-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802674

  • 329.85€ / 150µL

Expédition estimée
16-06-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody recognizes endogenous KRAP, which migrates with an apparent molecular mass of 180 kDa in SDS-PAGE.

SSFA2 gene ontology annotations related to this gene include actin binding.
Spécification

Spécification

KRAP/SSFA2
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P28290, Q922B9
Cleavage signal 1 protein, CS 1, CS1, KIAA1927, KRAP, SPAG13, sperm specific antigen 2, SSFA2
Rabbit
IgG
150 μL
-20°C
Primary
311146, 6744, 70599
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SSFA2
Liquid
Ssfa2
KRAP/SSFA2 Fusion Protein Ag5339
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents