missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Armoire de stockage standard Protector™

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

Marque:  Labconco™ 9900800

Code nomenclature Nacres: JA.01

Informations supplémentaires : Poids : 45.45000kg

 Afficher plus de versions de ce produit

Code produit. 15119715

  • 1094.44€ / 1 pièce


Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier

Includes: Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in. .

Description

Description

  • Support loads up to 800 lb. (363kg)
  • Durable epoxy-coated steel construction
  • Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
  • Manual-closing, non-locking door(s)

Order Info

Work surface and interior shelf kits are sold separately

Spécification

Spécification

55.88cm
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
Epoxy-Coated Steel
29.48kg
Single, Left Hinge, Manual Close
90.17 to 93.34cm
362.8kg
Storage Cabinet
30.48cm
Vidéos