missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Accessoires pour stations de ventilation Protector XVS™ : Ventilateurs à distance

Remote Blower for connection to XPert Balance Enclosure, Protector XVS Ventilation Station, Purifier HEPA Filtered Enclosure, Protector Work Station or Protector Demonstration Hood.

Marque:  Labconco™ 4863501

Code nomenclature Nacres: HA.23

Informations supplémentaires : Poids : 14.09000kg

 Afficher plus de versions de ce produit

Code produit. 15139725

  • 1187.18€ / 1 pièce


Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier

Description

Description

This Remote Blower is for connection to XPert Balance Enclosure, Protector XVS Ventilation Station, Purifier HEPA Filtered Enclosure, Protector Work Station or Protector Demonstration Hood. The direct drive blower has a 1/4 hp TEFC-type motor and overcomes external static pressure of 0.5 in. at 420 CFM. Housing and impeller are corrosion-resistant, phenolic-coated steel. Recommended for low to moderately corrosive conditions. Blower inlet is 5.94 in. OD and is sized to accept 6 in. nominal diameter PVC duct. Blower outlet is 4.25 in. x 7.4 in. OD.
Spécification

Spécification

5.6/2.8A
115/230V
Blower
50Hz
Epoxy-Coated Steel
Vidéos