missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Inserts pour tube de culture SteamScrubber™

Culture Tube Inserts for use with Labconco FlaskScrubber and SteamScrubber Glassware Washers.

Marque:  Labconco™ 4402101

Code nomenclature Nacres: NB.86

Informations supplémentaires : Poids : 1.36000kg

 Afficher plus de versions de ce produit

Code produit. 15129665

  • 198.48€ / 1 pièce


Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier

Description

Description

Type 304 stainless steel basket-style Inserts fit in standard open racks and spindle racks in SteamScrubber and FlaskScrubber Glassware Washers. Holds 105 each 20 to 24 mm OD tubes. Grid size is 26.5 mm x 26.5 mm. For use in lower rack only.
Spécification

Spécification

105 x 20 to 24mm O.D. tube
Each
Type 304 Stainless Steel
Culture Tube Insert
45.7cm
21.08cm
FlaskScrubber™, SteamScrubber™ glassware washer
25.14cm
1.1kg
Vidéos