missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27823-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Lysosomes are membrane-bound vacuoles which play a critical role in cellular metabolism. These organelles participate in endocytosis, the synthesis and assembly of acid hydrolases, act as sites for digestion of foreign materials and for specialized autolytic cellular processes. Membrane-associated glycoproteins have an important role in normal lysosomal function. Prior to posttranslational modification, Lysosome Associated Membrane Protein 2 (LAMP 2) is an approximately 45 kDa polypeptide. Mature, functional LAMP 2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of approximately 100-110 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail.Spécification
LAMP2 | |
Polyclonal | |
Unconjugated | |
LAMP2 | |
CD107b, LAMP 2, LAMP2, LAMPB, LGP110 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3920 | |
-20°C | |
Liquid |
Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13473 | |
LAMP2 | |
LAMP2 Fusion Protein Ag27183 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |