missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ LAPTM4B Recombinant Protein

sur devis

Spécification

Réactivité croisée Human
Formule 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Poids moléculaire 51
Nom LAPTM4B (Human) Recombinant Protein (P01)
Plage de pH 8
Consulter d'autres spécifications

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Spécification

Spécification

Human
51
8
Glutathione Sepharose 4 Fast Flow
Yes
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
LAPTM4B (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Vidéos
FDS
Documents

Documents

Product Certifications