missing translation for 'onlineSavingsMsg'
Learn More

LAPTM4B Recombinant Protein, Abnova

Marque:  Abnova H00055353-P01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16121463

  • 448.00€ / 25µg

Expédition estimée
22-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Spécification

Spécification

AAH14129
Solution
55353
LAPTM4B (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LAPTM4B
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
51
8
Glutathione Sepharose 4 Fast Flow
25 ug
MKMVAPWTRFYSNSCCLCCHVRTGTILLGVWYLIINAVVLLILLSALADPDQYNFSSSELGGVFEFMDDANMCIAIAISLLMILICAMATYGAYKQRAAWIIPFFCYQIFDFALNMLVAITVLIYPNSIQEYTRQLPPNFPYRDDVMSVNPTCLVLIILLFISIILTFKGYLISCVWNCYRYINGRNSSDVLVYVTSNDTTVLLPPYDDATVNGAAKEPPPPYVSA
LAPTM4beta|LC27
LAPTM4B
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents