missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22441-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli working through G protein-coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein has been observed to form a myeloid/lymphoid fusion partner in acute myeloid leukemia. Three transcript variants encoding different isoforms have been found for this gene.Spécification
LARG | |
Polyclonal | |
Unconjugated | |
ARHGEF12 | |
ARHGEF12, KIAA0382, LARG, Leukemia associated RhoGEF, PRO2792 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23365 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NZN5 | |
ARHGEF12 | |
LARG Fusion Protein Ag18211 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |